Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Tyrosine disodium salt dihydrate ≥98%, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Disodium 2-Amino-3-(4-oxidophenyl) propanoateL-3-(4-Hydroxyphenyl)alanine disodium salt

BAKR2094-05 2094-07 BAKR2094-05 BAKR2094-07
JT2094-5EA 434.8 USD InStock
CAJT2094-05 CAJT2094-7 JT2094-5 JT2094-7
L-Tyrosine disodium salt dihydrate ≥98%, BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
L(-)-Tyrosine disodium salt


Formula: C₉H₉NO₃Na₂·2H₂O
MW: 261.9 g/mol
Melting Pt: 200 °C
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD03840558
CAS Number: 122666-87-9
EINECS: 200-198-0
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₉H₉NO₃Na₂.2H₂O) ≥98 %
Appearance (white or tan crystals with an occasional brown crystal(s) Passes Test
Identification Passes Test
Endotoxin Concentration, IU/mg, For Information Only
Heavy Metals (as Pb) ≤15 ppm
Solution (10%) in Water Passes Test
pH of 1% Aqueous Solution at 25°C 10.5 - 11.5
Specific Rotation [Å]²º^D -14.4 - -12.6 Degree
Water (by KF) (H₂O) 12.0 - 16.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty