Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Tyrosine disodium salt dihydrate ≥99% (by HPLC), Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Disodium 2-Amino-3-(4-oxidophenyl) propanoateL-3-(4-Hydroxyphenyl)alanine disodium salt

Management of Change (MOC) category = R

BAKR2098-07 BAKR2098-07 BAKR2098-01 BAKR2098-06 BAKR2098-01
JT2098-06EA 1552.17 USD InStock
CAJT2098-01 JT2098-06 JT2098-07 CAJT2098-07 JT2098-01
L-Tyrosine disodium salt dihydrate ≥99% (by HPLC), Biotech Reagent, J.T.Baker®
L(-)-Tyrosine disodium salt


Formula: C₉H₉NO₃Na₂·2H₂O
MW: 261.9 g/mol
MDL Number: MFCD03840558
CAS Number: 122666-87-9
Order Now
Specification Test Results
Appearance (White to beige powder with occasional brown solid) Passes Test
Identification (IR) Passes Test
Solution (5%) in water Passes Test
Specific Rotation @ 20 °C °
Assay (by HPLC) ≥ 99 %
Water (by KF) ≤ 19.3 %
Bacterial endotoxins, EU/g ≤ 50
Arsenic (As), For Information Only ppm
Cadmium (Cd), For Information Only ppm
Mercury (Hg), For Information Only ppm
Cobalt (Co), For Information Only ppm
Nickel (Ni), For Information Only ppm
Vanadium (V), For Information Only ppm
Silver (Ag), For Information Only ppm
Iridium (Ir), For Information Only ppm
Chromium (Cr), For Information Only ppm
Copper (Cu), For Information Only ppm
Iron (Fe), For Information Only ppm
Lithium (Li), For Information Only ppm
Molybdenum (Mo), For Information Only ppm
Antimony (Sb), For Information Only ppm
Tin (Sn), For Information Only ppm
Aluminum (Al), For Information Only ppm
Manganese (Mn), For Information Only ppm
Residual Solvents: Class 3, each ≤ 5000 ppm
Total Aerobic Microbial Count ≤ 100 CFU/g
Total Yeasts and Molds ≤ 100 CFU/g
pH of 1% Solution, For Information Only
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Residual Solvents: Only Class 3 solvents are present as a result of use in the purification process. The concentration of Class 3 solvents is determined and reported for each production batch.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty