Specification Test Results
Meets E.P. Chemical Specifications |
|
Meets U.S.P Requirements |
|
Food GMP Manufactured Product |
|
Meets B.P. Chemical Specifications |
|
Meets F.C.C. Requirements |
|
GMP Manufactured Product |
|
CAUTION: For Manufacturing, processing or repackaging |
|
Bulk Food Chemical |
|
Bulk Pharmaceutical Chemical |
|
USP - Identification A |
Passes Test |
USP - Identification B - Diethylene Glycol |
≤0.10 % |
USP - Identification B - Ethylene Glycol |
≤0.10 % |
USP - Identification C |
Passes Test |
USP - Fatty Acids and Esters |
Passes Test |
USP - Specific Gravity at 25°/25°C |
≥1.249 |
USP - Color |
Passes Test |
USP - Chloride (Cl) |
≤0.001 % |
USP - Sulfate (SO₄) |
≤0.002 % |
USP - Chlorinated Compounds (as Cl) |
≤0.003 % |
USP - Assay (calculated on anhydrous basis) |
99.0 - 101.0 % |
USP - Water (by KF) (H₂O) |
≤5.0 % |
USP - Residue on Ignition |
≤0.01 % |
USP - Related Compounds: Individual Impurity |
≤0.1 % |
USP - Related Compo unds: Total Impurities |
≤1.0 % |
FCC - Identification A |
Passes Test |
FCC - Identification B (FCC) |
Passes Test |
FCC - Specific Gravity (FCC) |
≥1.259 |
FCC - Color |
Passes Test |
FCC - Residue on Ignition |
≤0.01 % |
FCC - Chlorinated Compounds (as Cl) |
≤0.003 % |
FCC - Fatty Acids and Esters(as butyric acid) |
Passes Test |
FCC - Readily Carbonizable Substances |
Passes Test |
FCC - Assay (as is) (FCC) |
99.0 - 101.0 % |
FCC - Water (by KF) (H₂O) (FCC) |
≤1.0 % |
FCC - Lead (Pb) |
≤1 mg/kg |
EP/BP - Appearance of Solution |
Passes Test |
EP/BP - Acidity or Alkalinity |
Passes Test |
EP/BP - Refractive Index, n²ºD |
1.470 - 1.475 |
EP/BP - Aldehydes |
≤10 ppm |
EP/BP - Ester |
Passes Test |
EP/BP - Halogenated Compounds |
≤35 ppm |
EP/BP - Sugars |
Passes Test |
EP/BP - Chloride (Cl) |
≤10 ppm |
EP/BP - Water (H₂O) |
≤2.0 % |
EP/BP - Ash (sulfated) |
≤0.01 % |
EP/BP - Impurity A and Related Substances - Impurity A |
≤0.1 % |
EP/BP - Impurity A and Related Substances - Other Impurities with R^T<Glycerol |
≤0.1 % |
EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol |
≤0.5 % |
EP - Assay (anhydrous basis) |
98.0 - 101.0 % |
EP/BP - Identification A |
Passes Test |
EP/BP - Identification B |
Passes Test |
Bacterial Endotoxin (EU/mg) |
≤0.05 |
No Class 1,2,3 or other solvents are used or produced in the |
|
manufacturing or purification of the product. |
|
Elemental Impurities (USP 232, EP 5.20) - Information on elemental |
|
impurities for this product is available on the associated Product |
|
Regulatory Data Sheet and elemental impurity profile report. |
|