Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

Management of Change (MOC) category = R

BAKR2140-QM BAKR2140-01 BAKR2140-01 BAKR2140-03 BAKR2140-R BAKR2140-QM BAKR2140-R BAKR2140-03
JT2140-3CS 5585.3 USD InStock
CAJT2140-R CAJT2140-3 JT2140-3 CAJT2140-QM JT2140-R JT2140-1 JT2140-QM CAJT2140-1
Glycerine 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Glycerine
Ideal for biotechnology, cell biology and molecular biology discovery, and biopharmaceutical production.

  • Food GMP manufactured product


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Food GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets F.C.C. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B - Diethylene Glycol ≤0.10 %
USP - Identification B - Ethylene Glycol ≤0.10 %
USP - Identification C Passes Test
USP - Fatty Acids and Esters Passes Test
USP - Specific Gravity at 25°/25°C ≥1.249
USP - Color Passes Test
USP - Chloride (Cl) ≤0.001 %
USP - Sulfate (SO₄) ≤0.002 %
USP - Chlorinated Compounds (as Cl) ≤0.003 %
USP - Assay (calculated on anhydrous basis) 99.0 - 101.0 %
USP - Water (by KF) (H₂O) ≤5.0 %
USP - Residue on Ignition ≤0.01 %
USP - Related Compounds: Individual Impurity ≤0.1 %
USP - Related Compo unds: Total Impurities ≤1.0 %
FCC - Identification A Passes Test
FCC - Identification B (FCC) Passes Test
FCC - Specific Gravity (FCC) ≥1.259
FCC - Color Passes Test
FCC - Residue on Ignition ≤0.01 %
FCC - Chlorinated Compounds (as Cl) ≤0.003 %
FCC - Fatty Acids and Esters(as butyric acid) Passes Test
FCC - Readily Carbonizable Substances Passes Test
FCC - Assay (as is) (FCC) 99.0 - 101.0 %
FCC - Water (by KF) (H₂O) (FCC) ≤1.0 %
FCC - Lead (Pb) ≤1 mg/kg
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Refractive Index, n²ºD 1.470 - 1.475
EP/BP - Aldehydes ≤10 ppm
EP/BP - Ester Passes Test
EP/BP - Halogenated Compounds ≤35 ppm
EP/BP - Sugars Passes Test
EP/BP - Chloride (Cl) ≤10 ppm
EP/BP - Water (H₂O) ≤2.0 %
EP/BP - Ash (sulfated) ≤0.01 %
EP/BP - Impurity A and Related Substances - Impurity A ≤0.1 %
EP/BP - Impurity A and Related Substances - Other Impurities with R^T<Glycerol ≤0.1 %
EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol ≤0.5 %
EP - Assay (anhydrous basis) 98.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
Bacterial Endotoxin (EU/mg) ≤0.05
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty