Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: GlysaninGlycerinGlyceritolPropane-1,2,3-triolOsmoglynPropanetriolGlycerolGlyrolGlycyl alcohol1,2,3-PropanetriolTrihydroxypropane

Management of Change (MOC) category = R

J.T. Baker®
BAKR2143-30 BAKR2143-07 BAKR2143-09 BAKR2143-09 BAKR2143-03 BAKR2143-01 BAKR2143-30 BAKR2143-03 BAKR2143-01 BAKR2143-07
JT2143-3CS 10296.24 USD InStock
CAJT2143-30 JT2143-3 CAJT2143-1 JT2143-1 CAJT2143-3 JT2143-30 CAJT2143-7 CAJT2143-9 JT2143-9 JT2143-7
Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Glycerine
Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives.


Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg
Order Now
Specification Test Results
Meets J.P. Chemical Specifications
GMP Manufactured Product
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B - Diethylene Glycol ≤0.10 %
USP - Identification B - Ethylene Glycol ≤0.10 %
USP - Identification C Passes Test
USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis) 99.0 - 101.0 %
USP - Specific Gravity at 25°/25°C ≥1.249
USP - Color Passes Test
USP - Residue on Ignition ≤0.01 %
USP - Chloride (Cl) ≤0.001 %
USP - Sulfate (SO₄) ≤0.002 %
USP - Chlorinated Compounds (as Cl) ≤0.003 %
USP - Fatty Acids and Esters Passes Test
USP - Water (H₂O) ≤5.0 %
USP - Related Compounds: Individual Impurity ≤0.1 %
USP - Related Compo unds: Total Impurities ≤1.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Refractive Index, n²ºD 1.470 - 1.475
EP/BP - Aldehydes ≤10 ppm
EP/BP - Ester Passes Test
EP/BP - Halogenated Compounds ≤35 ppm
EP/BP - Sugars Passes Test
EP/BP - Chloride (Cl) ≤10 ppm
EP/BP - Water (H₂O) ≤2.0 %
EP/BP - Ash (sulfated) ≤0.01 %
EP/BP - Impurity A and Related Substances - Impurity A ≤0.1 %
EP/BP - Impurity A and Related Substances - Other Impurities with R^T<Glycerol ≤0.1 %
EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol ≤0.5 %
EP - Assay (anhydrous basis) 98.0 - 101.0 %
JP - Assay (anhydrous basis) 98.0 - 101.0 %
JP - Identification Passes Test
JP - Specific Gravity at 20°/20°C ≥1.258
JP - Refractive Index, n²ºD ≥1.470
JP - Color Passes Test
JP - Residue on Ignition ≤0.01 %
JP - Chloride (Cl) ≤0.001 %
JP - Sulfate (SO₄) ≤0.002 %
JP - Heavy Metals (as Pb) ≤5 ppm
JP - Fatty Acids and Esters(as butyric acid) Passes Test
JP - Readily Carbonizable Substances Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Acrolein, Glucose, and Other Reducing Substances Passes Test
JP - Ammonium (NH₄) Passes Test
JP - Calcium (Ca) Passes Test
JP - Arsenic (As) ≤2 ppm
JP - Water (H₂O) ≤2.0 %
JP - Ethylene Glycol ≤0.1 %
JP - Diethylene Glycol ≤0.1 %
JP - Any Other Impurity ≤0.1 %
JP - Total Impurities ≤1.0 %
Endotoxin Concentration (EU/g) ≤5
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty