Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium 1-hexanesulfonate

Management of Change (MOC) category = N

BAKR2175-01 BAKR2175-05 BAKR2175-05 BAKR2175-01
JT2175-1CS 14907.21 USD InStock
CAJT2175-1 CAJT2175-5 JT2175-1 JT2175-5
1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
1-Hexanesulphonic acid sodium salt
Crystals. For ion-pair chromatography of basic compounds.


Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/94/2300794.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (as C₆H₁₃SO₃Na) (by acidimetry) ≥98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200
Appearance (clear, colorless, mobile liquid)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty