Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = N

BAKR2362-01 BAKR2362-01
JT2362-1CS 1306.46 USD InStock
JT2362-1 CAJT2362-1
Lithium carbonate ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Lithium carbonate
Powder. Lot analysis on label.


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Li₂CO₃) (by acidimetry) ≥99.0 %
Insoluble in Dilute HCl ≤0.01 %
Chloride (Cl) ≤0.005 %
Total Sulfur (as SO₄) ≤0.2 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.002 %
Calcium (Ca) ≤0.01 %
Potassium (K) ≤0.005 %
Silicon (Si), For Information Only
Sodium (Na) ≤0.010 %
Nitrate (NO₃) ≤5 ppm
Specific Surface Area, m²/g, For Information Only
Bulk Density (g/cc), For Information Only
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty