Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium acetate tetrahydrate 98.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Magnesium di(acetate) tetrahydrateAcetic acid magnesium salt tetrahydrate

Management of Change (MOC) category = N

BAKR2424-01 BAKR2424-05 BAKR2424-01 BAKR2424-05
JT2424-1CS 1865.03 USD InStock
CAJT2424-1 CAJT2424-5 JT2424-1 JT2424-5
Magnesium acetate tetrahydrate 98.0-102.0%, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium acetate tetrahydrate
Crystals. Lot analysis on label.


Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃COO)₂Mg·4H₂O) 98.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.5 - 8.5
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Barium (Ba) ≤0.001 %
Calcium (Ca) ≤0.01 %
Manganese (Mn) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Appearance (colorless, deliquescent solid)
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty