Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Magnesium dichloride

Management of Change (MOC) category = HR

BAKR2448-05 BAKR2448-07 BAKR2448-R BAKR2448-01 BAKR2448-07 BAKR2448-R BAKR2448-05 BAKR2448-01
JT2448-5CS 5084.39 USD InStock
CAJT2448-1 JT2448-5 CAJT2448-R JT2448-7 CAJT2448-5 CAJT2448-7 JT2448-R JT2448-1
Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®
Magnesium chloride hexahydrate
  • Food GMP manufactured product


Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684
Order Now
Specification Test Results
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 4.5 - 7.0
USP - Insoluble Matter ≤0.005 %
USP - Sulfate (SO₄) ≤0.005 %
USP - Barium (Ba) Passes Test
USP - Calcium (Ca) ≤0.01 %
USP - Potassium (K) Passes Test
FCC - Assay 99.0 - 101.0 %
FCC - Ammonium (NH₄) ≤0.005 %
FCC - Sulfate (SO₄) ≤0.005 %
FCC - Lead (Pb) ≤4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty