Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium sulfate, anhydrous ≥99%, powder, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Magnesium(II) sulfateBitter saltSulfuric acid magnesium salt (1:1)Epsom salt

Management of Change (MOC) category = N

BAKR2506-05 BAKR2506-05 BAKR2506-R BAKR2506-01 BAKR2506-01 BAKR2506-R
JT2506-1CS 841.07 USD InStock
JT2506-1 CAJT2506-5 CAJT2506-R CAJT2506-1 JT2506-5 JT2506-R
Magnesium sulfate, anhydrous ≥99%, powder, BAKER ANALYZED®, J.T.Baker®
Magnesium sulphate


Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (MgSO₄) (ignited basis) ≥99.0 %
Loss on Ignition ≤2.0 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.004 %
Ammonium (NH₄) ≤0.004 %
Calcium (Ca) ≤0.06 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Manganese (Mn) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty