Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(II) chloride tetrahydrate 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Manganese chloride tetrahydrateManganous chloride tetrahydrateManganese Chloride, TetrahydrateManganese(II) chloride tetrahydrateManganese dichloride tetrahydrateManganese chloride (tetrahydrate)

Management of Change (MOC) category = N

BAKR2540-05 BAKR2540-07 BAKR2540-01 BAKR2540-05 BAKR2540-R BAKR2540-01 BAKR2540-R BAKR2540-07
JT2540-7EA 1667.1 USD InStock
CAJT2540-7 JT2540-7 JT2540-5 JT2540-R JT2540-1 CAJT2540-1 CAJT2540-R CAJT2540-5
Manganese(II) chloride tetrahydrate 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Manganese (II) chloride tetrahydrate


Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 13,05751
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn) 98.0 - 101.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 3.5 - 6.0
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
ACS - Zinc (Zn) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty