Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, activated, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Molecular sieves pack 4ALinde 4A

Management of Change (MOC) category = N

BAKR2708-05 BAKR2708-01 BAKR2708-01
JT2708-1CS 627.54 USD InStock
CAJT2708-1 CAJT2708-5 JT2708-1
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, activated, J.T.Baker®
Molecular sieve A4 (0.4 nm, 4 Å)


MDL Number: MFCD03457537
CAS Number: 70955-01-0
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Loss on Drying at 150°C ≤5 %
Bead Size (mm) 1.6 - 2.5 mm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty