Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentanesulfonic acid sodium salt ≥98% for HPLC, for ion pair chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium pentane-1-sulfonateSodium pentanesulphonate, 98%Pentane-1-sulfonic acid sodium saltSodium pentane-1-sulphonateSodium pentanesulfonateSodium 1-pentanesulfonateSodium pentanesulfonate, 98%Pentane-1-sulphonic acid sodium saltSodium pentanesulphonateSodium 1-pentanesulphonate

Management of Change (MOC) category = N

BAKR2841-05 BAKR2841-05 BAKR2841-06 BAKR2841-06
JT2841-6EA 543.68 USD InStock
JT2841-6 JT2841-5 CAJT2841-5 CAJT2841-6
1-Pentanesulfonic acid sodium salt ≥98% for HPLC, for ion pair chromatography, J.T.Baker®
1-Pentanesulphonic acid sodium salt
Crystals. For ion-pair chromatography of basic compounds. UV cutoff 210nm.


Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/27/18570927.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (as C₅H₁₁SO₃Na) (by acidimetry) ≥98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty