Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperidine ≥99% (by GC), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: HexahydropyridineHexazanePentamethyleneimine

Management of Change (MOC) category = N

BAKR2895-05 BAKR2895-05 BAKR2895-98 BAKR2895-98
JT2895-05CS 497.36 USD InStock
JT2895-05 CAJT2895-98 JT2895-98 CAJT2895-5
Piperidine ≥99% (by GC), J.T.Baker®
Piperidine


Formula: C₅H₁₁N
MW: 85.15 g/mol
Density: 0.86 g/cm³
Flash Pt: 16.00 °C
MDL Number: MFCD00005979
CAS Number: 110-89-4
UN: 2401
ADR: 8,I
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₅H₁₁N) (by GC) ≥99 %
Water (H₂O) ≤0.2 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty