Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium acetate ≥99%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Diuretic salt4-Chloro-N-furfuryl-5-sulfamoylanthranlic acidAcetic acid potassium salt

Management of Change (MOC) category = NI

BAKR2912-05 BAKR2912-R BAKR2912-05 BAKR2912-R
JT2912-5CS 2903.46 USD InStock
CAJT2912-R JT2912-5 JT2912-R CAJT2912-5
Potassium acetate ≥99%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium acetate
The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.


Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 12,07764
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/72/16640572.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃COOK) (by non-aqueous titration) ≥99.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.5 - 9.0
Chloride (Cl) ≤0.003 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.03 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty