Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: KHPPotassium phthalate monobasicPotassium Acid PhthalatePotassium biphthalatePhthalic Acid Monopotassium SaltPhthalic acid potassium salt

Management of Change (MOC) category = N

BAKR2958-01 BAKR2958-05 BAKR2958-05 BAKR2958-00 BAKR2958-00 BAKR2958-07 BAKR2958-R BAKR2958-07 BAKR2958-01 BAKR2958-R
JT2958-0EA 176.3 USD InStock
CAJT2958-5 CAJT2958-7 JT2958-0 JT2958-1 JT2958-R JT2958-5 CAJT2958-0 CAJT2958-R JT2958-7 CAJT2958-1
Potassium hydrogen phthalate 99.95-100.05% (dry basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hydrogen phthalate
This reagent is satisfactory for use as a pH standard. For use as an acidimetric standard, this material should be lightly crushed and dried for 2 hours at 120 °C to remove any absorbed moisture.


Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 12,07773
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (1-KOCOC₆H₄-2-COOH) (dried basis) 99.95 - 100.05 %
Insoluble Matter ≤0.005 %
pH of 0.05M Solution at 25°C 4.00 - 4.02
Chlorinated Compounds (as Cl) ≤0.003 %
Sulfur Compounds (as S) ≤0.002 %
Sodium (Na) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
This reagent is satisfactory for use as a pH standard. For use as
anacidimetric standard, this material should be lightly crushed and dried for2 hours at 120° C to remove any absorbed moisture.
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty