Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen sulfate 90-100%, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potassium bisulphatePotassium bisulfate

Management of Change (MOC) category = N

BAKR2960-01 2960-01 BAKR2960-05 BAKR2960-05
JT2960-5CS 3703.19 USD InStock
JT2960-5 CAJT2960-1 CAJT2960-5 JT2960-1
Potassium hydrogen sulfate 90-100%, BAKER ANALYZED®, J.T.Baker®
Potassium hydrogen sulphate
Lot analysis on label.


Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 12,07774
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Acidity (as H₂SO₄) 35.0 - 37.0 %
Insoluble Matter and NH₄OH Precipitate ≤0.01 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Calcium and Magnesium (as Ca) (by EDTA titration) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty