Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium bromide ≥99.0%, BAKER INSTRA-ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = N

BAKR2961-03 BAKR2961-05 BAKR2961-03 BAKR2961-05
JT2961-3EA 294.86 USD InStock
JT2961-3 JT2961-5 CAJT2961-5 CAJT2961-3
Potassium bromide ≥99.0%, BAKER INSTRA-ANALYZED®, J.T.Baker®
Potassium bromide
For infrared analysis. Lot analysis on label.


Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 12,07780
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (KBr) (by Ag titrn) ≥99.0 %
Insoluble Matter ≤0.005 %
Loss on Drying at 105°C ≤0.05 %
pH of 5% Solution at 25°C 5.0 - 7.5
Nitrogen Compounds (as N) ≤0.0005 %
Sulfate (SO₄) ≤0.002 %
Suitability for Infrared Analysis Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty