Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate sesquihydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = N

BAKR3010-05 BAKR3010-05 BAKR3010-01 BAKR3010-R BAKR3010-01 BAKR3010-R
JT3010-5CS 2649.14 USD InStock
CAJT3010-5 CAJT3010-R CAJT3010-1 JT3010-5 JT3010-1 JT3010-R
Potassium carbonate sesquihydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium carbonate sesquihydrate


Formula: CK₂O₃·1.5H₂O
MW: 165.23 g/mol
Melting Pt: 891 °C
Density: 2.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₂CO₃·1.5H₂O) 98.5 - 101.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Loss on Heating at 285°C 14.0 - 16.5 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Magnesium (Mg) ≤0.002 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Silica (SiO₂) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.004 %
ACS - Ammonium Hydroxide Precipitate ≤0.01 %
ACS - Sodium (Na) ≤0.02 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty