Synonyms:
Potassiumchloride
Management of Change (MOC) category = N
J.T. Baker®
BAKR3040-01
3040-69
BAKR3040-R
BAKR3040-05
BAKR3040-19
3040-69
BAKR3040-01
BAKR3040-07
BAKR3040-19
BAKR3040-05
BAKR3040-09
BAKR3040-R
BAKR3040-07
BAKR3040-09
JT3040-1CS
434.8
USD
InStock
JT3040-1
JT3040-69
JT3040-9
CAJT3040-69
JT3040-7
JT3040-5
JT3040-R
CAJT3040-19
CAJT3040-9
CAJT3040-7
JT3040-19
CAJT3040-5
CAJT3040-R
CAJT3040-1
Potassium chloride 99.0-100.5% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium chloride