Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Potassiumchloride

Management of Change (MOC) category = R

BAKR3046-07 BAKR3046-01 BAKR3046-A1 BAKR3046-05 BAKR3046-01 BAKR3046-05 BAKR3046-A1 BAKR3046-07 BAKR3046-R BAKR3046-R
JT3046-A1EA 4429.5 USD InStock
CAJT3046-5 CAJT3046-A1 JT3046-A1 CAJT3046-R CAJT3046-1 JT3046-7 JT3046-5 JT3046-R JT3046-1 CAJT3046-7
Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Potassium chloride
Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the p


Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704
Order Now
Specification Test Results
GMP Manufactured Product
Meets U.S.P Requirements
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (KCl) (dried basis) 99.0 - 100.5 %
USP - Acidity or Alkalinity Passes Test
USP - Calcium and Magnesium Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Bromide (Br) ≤0.1 %
USP - Iodides (I) ≤0.005 %
USP - Loss on Drying ≤1.0 %
USP - Sodium (Na) Passes Test
FCC - Assay (KCl) (dried basis) 99.0 - 100.5 %
FCC - Acidity or Alkalinity Passes Test
FCC - Heavy Metals (as Pb) ≤5 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iodide and/or Bromide Passes Test
FCC - Loss on Drying ≤1.0 %
FCC - Sodium (Na) Passes Test
Appearance (White crystals) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty