Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

Management of Change (MOC) category = N

BAKR3115-01 BAKR3115-05
JT3115-1CS 310.12 USD InStock
JT3115-1 JT3115-5
Sodium hydroxide ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (NaOH) (by acidimetry) ≥98 %
Calcium (Ca) ≤0.005 %
Chloride (Cl) ≤0.005 %
Copper (Cu) ≤0.001 %
Heavy Metals (as Ag) ≤0.001 %
Insoluble Matter ≤0.003 %
Iron (Fe) ≤0.0003 %
ACS - Magnesium (Mg) ≤0.002 %
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.0003 %
Phosphate (PO₄) ≤0.0002 %
Potassium (K) ≤0.02 %
Sodium Carbonate (Na₂CO₃) ≤0.4 %
ACS - Sulfate (SO₄) ≤0.003 %
Meets Reagent Specifications for testing USP/NF monographs
Appearance (white hygroscopic pellets)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty