Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium iodate 99.4-100.4%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = N

BAKR3156-01 BAKR3156-04 BAKR3156-05
JT3156-4CS 727.74 USD InStock
CAJT3156-5 JT3156-4 JT3156-1
Potassium iodate 99.4-100.4%, BAKER ANALYZED® ACS, J.T.Baker®
Potassium iodate


Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560 °C
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 12,07808
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KIO₃) 99.4 - 100.4 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 5.0 - 8.0
Chloride and Bromide (as Cl) ≤0.01 %
Iodide (I) ≤0.001 %
Nitrogen Compounds (as N) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty