Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium nitrate 99.0-100.5%, granular USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Management of Change (MOC) category = HR

BAKR3193-85 BAKR3193-85
JT3193-85CS 1171.84 USD InStock
JT3193-85 CAJT3193-85
Potassium nitrate 99.0-100.5%, granular USP, Multi-Compendial, J.T.Baker®
Potassium nitrate


Formula: KNO₃
MW: 101.1 g/mol
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 12,07815
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets U.S.P Requirements
Meets F.C.C. Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (KNO₃) 99.0 - 100.5 %
USP - Arsenic (As) ≤3 ppm
USP - Chloride (Cl) ≤0.03 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iron (Fe) ≤0.001 %
USP - Lead (Pb) ≤4 mg/kg
USP - Nitrite (NO₂) ≤5 ppm
USP - Sodium (Na) ≤0.1 %
USP - Sulfate (SO₄) ≤0.1 %
JSFA - Arsenic (As) ≤2 ppm
JSFA - Chloride (Cl) ≤0.021 %
JSFA - Clarity and Color of Solution Passes Test
JSFA - Loss on Drying at 105°C (4 hours) ≤1 %
EP/BP - Assay (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Reducing Substances Passes Test
EP/BP - Chloride (Cl) ≤20 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Ammonium (NH₄) ≤100 ppm
EP/BP - Calcium (Ca) ≤100 ppm
EP/BP - Iron (Fe) ≤20 ppm
EP/BP - Sodium (Na) ≤0.10 %
EP/BP - Loss on Drying at 105°C ≤0.5 %
FCC - Assay (KNO₃) (dried basis) 99.0 - 100.5 %
FCC - Chlorate Passes Test
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying at 105°C (4 hours) ≤1 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
STORAGE: Preserve in tight containers.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty