Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium oxalate monohydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Potassium oxalate monohydratePotassium ethanedioate monohydrateOxalic acid potassium salt monohydratePotassium oxalate hydrate

Management of Change (MOC) category = N

BAKR3212-05 BAKR3212-01 BAKR3212-01
JT3212-5CS 4101.67 USD InStock
JT3212-5 CAJT3212-1 JT3212-1
di-Potassium oxalate monohydrate 98.5-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
di-Potassium oxalate monohydrate


Formula: (COOK)₂·H₂O
MW: 184.23 g/mol
Melting Pt: 356 °C
Density: 2.154 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III
Merck Index: 12,07820
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (K₂C₂O₄·H₂O) 98.5 - 101.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Neutrality Passes Test
ACS - pH of 5% Solution at 25°C 7.0 - 8.5
ACS - Chloride (Cl) ≤0.002 %
ACS - Sulfate (SO₄) ≤0.01 %
ACS - Ammonium (NH₄) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.001 %
ACS - Sodium (Na) ≤0.02 %
ACS - Substances Darkened by Hot H₂SO₄ Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty