Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Potassium metaperiodatePotassium tetroxoiodate (VII)

Management of Change (MOC) category = N

BAKR3224-04 BAKR3224-01 BAKR3224-04 BAKR3224-01
JT3224-4CS 3609.33 USD InStock
JT3224-4 CAJT3224-1 CAJT3224-4 JT3224-1
Potassium periodate 99.8-100.3% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Potassium periodate


Formula: KlO₄
MW: 230 g/mol
Melting Pt: 576…588 °C
Density: 3.62 g/cm³ (25 °C)
MDL Number: MFCD00011407
CAS Number: 7790-21-8
EINECS: 232-196-0
UN: 1479
ADR: 5.1,II
Merck Index: 12,07823
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KIO₄) 99.8 - 100.3 %
Insoluble Matter ≤0.01 %
Other Halogens (as Cl) ≤0.01 %
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty