Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKR3227-01 BAKR3227-05
JT3227-1CS 660.52 USD InStock
JT3227-1 JT3227-5
Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS for the determination of mercury, J.T.Baker®
Potassium permanganate


Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: >240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KMnO₄) ≥99.0 %
ACS - Insoluble Matter ≤0.2 %
ACS - Chloride and Chlorate (as Cl) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.02 %
Trace Impurities - Mercury (Hg) ≤0.050 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty