Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate ≥98.0% (by acidimetry), powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: di-Potassium hydrogen orthophosphateDipotassium hydrogenphosphateDibasic Potassium PhosphatePotassium phosphate dibasicPotassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)Potassium hydrogen phosphateDipotassium hydrogen phosphateDipotassium PhosphateDipotassium phosphate

Management of Change (MOC) category = N

J.T. Baker®
BAKR3252-R BAKR3252-01 BAKR3252-05 3252-07 BAKR3252-05 BAKR3252-07 BAKR3252-01 BAKR3252-R
JT3252-7EA 1470.4 USD InStock
CAJT3252-R CAJT3252-1 JT3252-7 CAJT3252-7 JT3252-01 JT3252-15 CAJT3252-5 JT3252-R
di-Potassium hydrogen phosphate ≥98.0% (by acidimetry), powder, BAKER ANALYZED® ACS, J.T.Baker®
di-Potassium hydrogen phosphate


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 12,07828
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (K₂HPO₄) (by acidimetry) ≥ 98.0 %
Insoluble Matter ≤ 0.01 %
Loss on Drying at 105°C ≤ 1.0 %
pH of 5% Solution at 25°C 8.5 - 9.6
Chloride (Cl) ≤ 0.003 %
Fluoride (F) ≤ 0.001 %
Nitrogen Compounds (as N) ≤ 0.001 %
Sulfate (SO₄) ≤ 0.005 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Sodium (Na) ≤ 0.05 %
Trace Impurities - Arsenic (As) ≤ 1 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Lead (Pb) ≤ 5 ppm
Color (APHA), For Information Only
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty