Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: di-Potassium hydrogen orthophosphateDibasic Potassium PhosphatePotassium phosphate dibasicPotassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)Potassium hydrogen phosphateDipotassium hydrogen phosphate

Management of Change (MOC) category = RL

BAKR3254-05 BAKR3254-01 BAKR3254-05 BAKR3254-01
JT3254-5CS 3256.74 USD InStock
CAJT3254-1 JT3254-5 CAJT3254-5 JT3254-1
di-Potassium hydrogen phosphate ≥98.0% (dried basis) FCC, J.T.Baker®
di-Potassium hydrogen phosphate
  • Food GMP manufactured product


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 12,07828
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now
Specification Test Results
Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Identification Passes Test
FCC - Assay (K₂HPO₄) (dried basis) ≥98.0 %
FCC - Insoluble Substances ≤0.2 %
FCC - Loss on Drying ≤2.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Fluoride (F) ≤10 ppm
FCC - Lead (Pb) ≤2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty