Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrateRochelle saltL(+)-Tartaric acid potassium sodium salt tetrahydratePotassium sodium tartrate tetrahydratePotassium sodium L(+)-tartrate tetrahydrate(+)-Potassium sodium L-tartrate tetrahydrateSodium potassium tartratePotassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate(R,R)-(+)-Potassium sodium tartrate tetrahydrate

J.T. Baker®
BAKR3262-07 BAKR3262-07 BAKR3262-05 BAKR3262-19 BAKR3262-01 BAKR3262-05 BAKR3262-01 BAKR3262-19
JT3262-01CS 1407.41 USD InStock
CAJT3262-7 JT3262-01 JT3262-05 CAJT3262-5 CAJT3262-19 JT3262-19 CAJT3262-1 JT3262-7
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
L(+)-Potassium sodium tartrate tetrahydrate


Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: 1.79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 12,07840
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KOCO(CHOH)₂COONa·4H₂O) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.0 - 8.5
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Ammonium (NH₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty