Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 99.0 - 100.5% 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: HaliteSea saltSaline

BAKR3302-25 3302-28 BAKR3302-25 BAKR3302-28
JT3302-28EA 450.3 USD InStock
CAJT3302-28 CAJT3302-25 JT3302-28 JT3302-25
Sodium chloride 99.0 - 100.5% 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Melting Pt: 801 °C
Density: 2.16 g/ml (25 °C)
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now
Specification Test Results
GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Appearance of Solution Passes Test
USP - Aluminum (Al) ≤0.2 ppm
USP - Arsenic (As) ≤1 ppm
USP - Barium (Ba) Passes Test
USP - Bromide (Br) ≤100 ppm
USP - Ferrocyanide Passes Test
USP - Iodides (I) Passes Test
USP - Iron (Fe) ≤2 ppm
USP - Loss on Drying at 105°C ≤0.5 %
USP - Magnesium and Alkaline Earth Metals (as Ca) ≤100 ppm
USP - Nitrite (NO₂) (Absorbance Value) ≤0.01
USP - Phosphate (PO₄) ≤25 ppm
USP - Potassium (K) ≤500 ppm
USP - Sulfate (SO₄) ≤200 ppm
USP - Assay (NaCl) (dried basis) 99.0 - 100.5 %
FCC - Assay (as NaCl) (FCC) (ignited basis) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Arsenic (As) ≤1 ppm
FCC - Calcium and Magnesium ≤0.35 %
FCC - Heavy Metals (as Pb) ≤2 ppm
FCC - Loss on Drying at 105°C ≤0.5 %
Solution Test Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Must be subjected to further processing during the preparation of
injectable dosage forms
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty