Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium thiocyanate ≥98.5% (Ag basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Potassium rhodanide

Management of Change (MOC) category = NI

BAKR3326-R BAKR3326-07 BAKR3326-R
JT3326-7EA 3079.37 USD InStock
JT3326-7 CAJT3326-R JT3326-R
Potassium thiocyanate ≥98.5% (Ag basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium thiocyanate
Crystals. Lot analysis on label.


Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (KSCN) (by Ag titrn) ≥98.5 %
Insoluble in H₂O ≤0.005 %
pH of 5% Solution at 25°C 5.3 - 8.7
Chloride (Cl) ≤0.005 %
Sulfate (SO₄) ≤0.005 %
Ammonium (NH₄) ≤0.003 %
Sodium (Na) ≤0.005 %
Iodine Consuming Substances (meq/g) ≤0.004
Use Test Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty