Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine ≥99.0% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

BAKR3348-98 BAKR3348-33 BAKR3348-22 BAKR3348-03 BAKR3348-33 BAKR3348-22 BAKR3348-01 BAKR3348-98 BAKR3348-01 BAKR3348-03
JT3348-03CS 10685.04 USD InStock
JT3348-03 CAJT3348-33 CAJT3348-22 JT3348-01 CAJT3348-98 JT3348-98 JT3348-33 JT3348-22 CAJT3348-1 CAJT3348-3
Pyridine ≥99.0% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Pyridine
Lot analysis on label.


Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.98 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 12,08153
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₅H₅N) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤20
Solubility in H₂O Passes Test
Residue after Evaporation ≤0.0020 %
Water (by KF, volumetric) ≤0.1 %
Sulfate (SO₄) ≤0.001 %
Ammonia (as NH₃) ≤0.002 %
Reducing Substances Passes Test
Chloride (Cl) ≤5 ppm
Trace Impurities - Copper (Cu) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty