Synonyms:
Sulphuric acid disodium salt
, Bisodium sulphate
, Sulfuric acid disodium salt
, Bisodium sulfate
, Disodium sulphate
, Disodium sulfate
Management of Change (MOC) category = N
BAKR3375-09
BAKR3375-05
BAKR3375-05
BAKR3375-09
BAKR3375-01
BAKR3375-01
BAKR3375-07
BAKR3375-07
JT3375-7EA
975.99
USD
InStock
CAJT3375-1
JT3375-7
JT3375-5
CAJT3375-5
JT3375-9
CAJT3375-9
CAJT3375-7
JT3375-1
Sodium sulfate, anhydrous ≥99.0%, granular, ULTRA RESI-ANALYZED® ACS 12-60 Mesh for pesticide residue analysis, J.T.Baker®
Sodium sulphate
BAKER ULTRA RESI-ANALYZED® Reagent ultra high purity solvents and salts developed for organic residue extraction/concentration procedures.