Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium chloride ≥99.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Ammonium muriateSalmiac

Management of Change (MOC) category = N

MACR3384-06 MACR3384-12 MACR3384-09 MACR3384-25 MACR3384-06 MACR3384-09 MACR3384-20 MACR3384-20 MACR3384-12 MACR3384-09
MK3384-09CS 1969.16 USD InStock
CAMK338420 MK3384-09 MK338420 MK338412 CAMK338406 MK3384-25 JT3384-09 MK338406 CAMK3384-09 CAMK338412
Ammonium chloride ≥99.5% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Ammonium chloride


Formula: NH₄Cl
MW: 53.49 g/mol
Melting Pt: 335 °C
Density: 1.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (NH₄Cl) (by Ag titrn) ≥99.5 %
ACS - Calcium (Ca) ≤0.001 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤2 ppm
ACS - Magnesium (Mg) ≤5 ppm
ACS - pH of 5% Solution at 25°C 4.5 - 5.5
ACS - Phosphate (PO₄) ≤2 ppm
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Appearance (white crystals) Passes Test
Heavy Metals (as Pb) ≤2 ppm
Nitrate (NO₃) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty