Management of Change (MOC) category = N
BAKR3401-01
BAKR3401-05
BAKR3401-05
JT3401-5CS
2809.25
USD
InStock
CAJT3401-5
JT3401-5
JT3401-1
Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®
Silica gel
6 to 18 mesh. Indicating (cobalt chloride). Lot analysis on label.
Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed