Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKR3402-05 BAKR3402-01 BAKR3402-05 BAKR3402-01
JT3402-5EA 661.12 USD InStock
CAJT3402-5 JT3402-5 CAJT3402-1 JT3402-1
Silica gel, BAKER ANALYZED® (4-10 Mesh) desiccant, with moisture indicator, J.T.Baker®
Silica gel
Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed


Formula: SiO₂
MW: 60.08 g/mol
Density: 2.10 kg/m³
MDL Number: MFCD00011232
CAS Number: 63231-67-4
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Loss on Heating at 200°C ≤6.0 %
Suitability for Moisture Absorption at 80% Relative Humidity ≥33 %
Suitability for Moisture Absorption at 20% Relative Humidity ≥10 %
Suitability as Indicating Desiccant Passes Test
Mesh - Thru U.S. No. 10 Sieve ≤5.0 %
Color Change: Blue.................Activated Violet..........10% Moisture Absorbed Pink............19% Moisture Absorbed Pale Pink.......28% Moisture Absorbed

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty