Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acidHydroxytricarballylic acid

Filtered through a 0.2 µm filter.

J.T. Baker®
BAKR0341-07 BAKR0341-07 BAKR0341-25 BAKR0341-27 BAKR0341-26 BAKR0341-26 BAKR0341-27
JT34107EA 1240.12 USD InStock
JT34107 JT0341-25 JT0341-26 CAJT0341-7 JT0341-27 CAJT0341-26 CAJT0341-27
Citric acid 50% in aqueous solution, Biotech Reagent, J.T.Baker®
Citric acid
Made from purified water USP and citric acid, anhydrous USP/ACS.


Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Density: 1.24 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg
Order Now
Specification Test Results
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Assay (HOC(COOH)(CH₂COOH)₂) 48 - 52 %
Identification Passes Test
Chloride (Cl) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤3 ppm
Phosphate (PO₄) ≤0.001 %
Made from Purified Water U.S.P. and Citric Acid, anhydrous U.S.P./A.C.S.
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty