Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

Management of Change (MOC) category = N

BAKR3426-04 BAKR3426-04 BAKR3426-01 BAKR3426-01 BAKR3426-00
JT3426-4CS 4853.2 USD InStock
CAJT3426-4 CAJT3426-1 JT3426-4 JT3426-1 JT3426-0
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Density: 4.35 g/cm³
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (AgNO₃) ≥99.7 %
Clarity of Solution Passes Test
Free Acid Passes Test
Substances Not Precipitated by HCl ≤0.01 %
Sulfate (SO₄) ≤0.002 %
Lead (Pb) ≤0.001 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Copper (Cu) ≤2.000 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty