Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Benzoic acid sodium salt

Management of Change (MOC) category = R

BAKR3501-20 3501-05 BAKR3501-20 BAKR3501-05 BAKR3501-20
JT3501-20EA 3083.22 USD InStock
CAJT3501-5 CAJT3501-20 JT3501-20 TXLJT350120ORE JT3501-5
Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Sodium benzoate


Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 12,08725
Order Now
Specification Test Results
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Identification C Passes Test
NF - Alkalinity Passes Test
NF - Water (H₂O) ≤ 1.5 %
NF - Assay (C₆H₅COONa) (anhydrous basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Halogenated Compounds (as Cl) - Ionized Chlorine ≤ 200 ppm
EP/BP - Halogenated Compounds (as Cl) - Total Chlorine ≤ 300 ppm
EP/BP - Loss on Drying ≤ 2.0 %
EP/BP - Assay(C₆H₅COONa)(dried basis) 99.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty