Synonyms:
Sodium bicarbonate
Management of Change (MOC) category = R
BAKR3509-07
BAKR3509-09
BAKR3509-05
BAKR3509-09
BAKR3509-05
BAKR3509-07
BAKR3509-01
BAKR3509-01
JT3509-1CS
317.74
USD
InStock
JT3509-1
JT3509-7
CAJT3509-1
JT3509-5
CAJT3509-5
JT3509-9
CAJT3509-9
CAJT3509-7
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Sodium hydrogen carbonate
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Certifications: Meets USP and FCC requirements.
Caution: Not for use in hemodialysis.