Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

BAKR3510-07 BAKR3510-05 BAKR3510-07 BAKR3510-09 BAKR3510-05 BAKR3510-09
JT3510-5CS 1203.43 USD InStock
CAJT3510-9 CAJT3510-5 CAJT3510-7 JT3510-5 JT3510-9 JT3510-7
Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
Sodium hydrogen carbonate
Powder. Pyrogen tested. GMP manufactured.


Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 12,08726
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now
Specification Test Results
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification Passes Test
USP - Limit of Sulfur Compounds ≤ 0.015 %
USP - Loss on Drying ≤ 0.25 %
USP - Insoluble Matter Passes Test
USP - Normal Carbonate Passes Test
USP - Limit of Ammonia ≤ 20 ppm
USP - Chloride (Cl) ≤ 0.015 %
USP - Arsenic (As) ≤ 2 ppm
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Assay (NaHCO₃) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Carbonate (CO₃) Passes Test
EP/BP - Chloride (Cl) ≤ 150 ppm
EP/BP - Sulfate (SO₄) ≤ 150 ppm
EP/BP - Ammonium (NH₄) ≤ 20 ppm
EP/BP - Calcium (Ca) ≤ 100 ppm
EP/BP - Iron (Fe) ≤ 20 ppm
JP - Assay (NaHCO₃) 99.0 - 101.0 %
JP - Identification Passes Test
JP - pH (1 in 20) 7.9 - 8.4
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.040 %
JP - Carbonate (CO₃) Passes Test
JP - Ammonium (NH₄) Passes Test
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Arsenic (As) ≤ 2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty