Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen sulfate monohydrate 35.0–36.5% (by acidimetry), crystals, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium bisulfate monohydrateSodium bisulphate monohydrate

Management of Change (MOC) category = N

BAKR3534-01 BAKR3534-01 BAKR3534-05 BAKR3534-05
JT3534-5CS 3112.39 USD InStock
CAJT3534-5 JT3534-5 CAJT3534-1 JT3534-1
Sodium hydrogen sulfate monohydrate 35.0–36.5% (by acidimetry), crystals, BAKER ANALYZED®, J.T.Baker®
Sodium hydrogen sulphate monohydrate
Crystals.


Formula: NaHSO₄·H₂O
MW: 138.08 g/mol
Melting Pt: 183 °C
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 12,08729
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (H₂SO₄) (by acidimetry) 35.0 - 36.5 %
Insoluble Matter and NH₄OH Precipitate ≤0.005 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Calcium and Magnesium (as Ca) (by EDTA titration) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty