Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium metabisulfite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Pyrosulfurous acid disodium saltDisodium disulfiteSodium disulphiteDisodium disulphiteSodium pyrosulphiteSodium disulfitedi-Sodium disulphiteSodium pyrosulfite

Management of Change (MOC) category = N

BAKR3552-07 BAKR3552-07 BAKR3552-01 BAKR3552-01 BAKR3552-05 BAKR3552-05
JT3552-1CS 324.77 USD InStock
JT3552-1 CAJT3552-7 CAJT3552-5 JT3552-5 CAJT3552-1 JT3552-7
Sodium metabisulfite ≥97.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Sodium metabisulphite


Formula: Na₂S₂O₅
MW: 190.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
Merck Index: 13,08712
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/32/16640632.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂S₂O₅) ≥97.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.05 %
Thiosulfate (S₂O₃) ≤0.05 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.002 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty