Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium tetraborate decahydrate 99.5-105.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium tetraborate decahydrateBorax tetra

Management of Change (MOC) category = N

BAKR3568-05 BAKR3568-07 BAKR3568-05 BAKR3568-07
JT3568-7EA 1034.87 USD InStock
CAJT3568-7 CAJT3568-5 JT3568-7 JT3568-5
di-Sodium tetraborate decahydrate 99.5-105.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
di-Sodium tetraborate decahydrate
Crystals. Lot analysis on label.


Formula: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Na₂B₄O₇·10H₂O) 99.5 - 105.0 %
Insoluble Matter ≤0.005 %
pH of 0.01M Solution at 25°C 9.15 - 9.20
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Calcium (Ca) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty