Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium tetraborate decahydrate 99.0-105.0% NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Sodium tetraborate decahydrateBorax tetra

Management of Change (MOC) category = R

BAKR3574-01 BAKR3574-05 BAKR3574-QM BAKR3574-05 BAKR3574-01 BAKR3574-07 BAKR3574-QM 3574-07
JT3574-1CS 1587.54 USD InStock
JT3574-1 CAJT3574-7 JT3574-QM CAJT3574-5 CAJT3574-QM CAJT3574-1 JT3574-7 JT3574-5
di-Sodium tetraborate decahydrate 99.0-105.0% NF, J.T.Baker®
di-Sodium tetraborate decahydrate
  • GMP manufactured product


Formula: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 75 °C
Density: 1.73 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662
Order Now
Specification Test Results
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay 99.0 - 105.0 %
NF - Identification Passes Test
NF - Carbonate and Bicarbonate Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty