Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals FCC, NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Sodium carbonate hydrousSoda monohydrate

Management of Change (MOC) category = R

BAKR3600-05 BAKR3600-S1 BAKR3600-09 BAKR3600-01 BAKR3600-D1 BAKR3600-09 BAKR3600-QM BAKR3600-S1 BAKR3600-05 BAKR3600-QM BAKR3600-S1 BAKR3600-D1 BAKR3600-01
JT3600-9EA 9203.05 USD InStock
JT3600-9 CAJT3600-5 JT3600-D1 CAJT3600-9 CAJT3600-D1 JT3600-1 CAJT3600-S1 JT3600-5 TXLJT3600S1CTT JT3600-QM JT3600-S1 CAJT3600-QM CAJT3600-1
Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals FCC, NF, J.T.Baker®
Sodium carbonate monohydrate


Formula: Na₂CO₃·1H₂O
MW: 124.00 g/mol
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6
EINECS: 207-838-8
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) 12.0 - 15.0 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤4 mg/kg
FCC - Loss on Drying 12.0 - 15.0 %
FCC - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty