Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Calcined sodaCarbonic acid disodium saltNa-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

Management of Change (MOC) category = R

BAKR3605-QM BAKR3605-QM BAKR3605-01 BAKR3605-R BAKR3605-09 3605-06 BAKR3605-R 3605-09 BAKR3605-06 BAKR3605-01
JT3605-6EA 668.16 USD InStock
JT3605-6 CAJT3605-R CAJT3605-QM CAJT3605-1 JT3605-9 CAJT3605-6 JT3605-QM CAJT3605-9 JT3605-R JT3605-1
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®
Sodium carbonate
pH adjusters are used to adjust the pH of formulations.

  • Food GMP manufactured product


Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) ≤0.5 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying ≤0.5 %
FCC - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Lead (Pb) ≤4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty