Synonyms:
HaliteRock saltSalineSaltSea salt
Management of Change (MOC) category = R
J.T. Baker®
BAKR3628-F9
BAKR3628-W2
BAKR3628-B7
BAKR3628-68
BAKR3628-W3
BAKR3628-S1
BAKR3628-01
BAKR3628-05
BAKR3628-08
BAKR3628-07
BAKR3628-A2
BAKR3628-05
BAKR3628-60
BAKR3628-F7
BAKR3628-60
BAKR3628-F9
BAKR3628-QM
BAKR3628-W2
BAKR3628-20
BAKR3628-68
BAKR3628-2Y
BAKR3628-SB
BAKR3628-06
BAKR3628-07
BAKR3628-A1
BAKR3628-01
BAKR3628-W3
BAKR3628-65
BAKR3628-B7
BAKR3628-08
BAKR3628-F7
BAKR3628-SB
BAKR3628-A1
BAKR3628-A2
BAKR3628-QM
BAKR3628-ZS
BAKR3628-06
BAKR3628-65
BAKR3628-20
JT3628-F7EA
321
USD
InStock
CAJT3628-SB
JT3628-F7
JT3628-F9
CAJT3628-A2
CAJT3628-A1
JT3628-B7
JT3628-2Y
JT362865
JT3628-S1
CAJT3628-QM
JT3628-20
JT3628-68
JT3628-ZS
JT3628-W2
JT3628-60
JT3628-W3
JT3628-SB
JT3628-5
JT3628-A2
JT3628-A1
CAJT3628-F7
JT3628-8
CAJT3628-F9
JT3628-6
CAJT3628-1
JT3628-7
CAJT3628-6
CAJT3628-7
CAJT3628-5
JT3628-1
CAJT3628-8
CAJT3628-W3
CAJT3628-65
CAJT3628-20
CAJT3628-68
CAJT3628-B7
CAJT3628-W2
CAJT3628-60
JT3628-QM
Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Sodium chloride
Certifications: Must be subjected to further processing during the preparation of injectable dosage forms. Meets USP and FCC requirements. GMP manufactured product.
Caution: Preserve in tightly sealed containers.