Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateTrisodium citrateSodium citrate dihydrateSodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrateCitric acid tridosium salt dihydrate

Management of Change (MOC) category = N

BAKR3646-R BAKR3646-01 BAKR3646-07 BAKR3646-05 BAKR3646-R BAKR3646-07 BAKR3646-01 BAKR3646-05
JT3646-7EA 1122.23 USD InStock
CAJT3646-R CAJT3646-1 JT3646-7 CAJT3646-7 CAJT3646-5 JT3646-1 JT3646-5 JT3646-R
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
tri-Sodium citrate dihydrate
  • Lot analysis on label


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C
Density: 1.70 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅Na₃O₇·2H₂O) (by non-aqueous titration) 99.0 - 101.0 %
ACS - pH of 5% Solution at 25°C 7.0 - 9.0
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Sulfate (SO₄) ≤0.005 %
ACS - Ammonia (as NH₃) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty